missing translation for 'onlineSavingsMsg'
Learn More

Air-Tite™ Sterile Syringes

Catalog No. 14817178
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
20 mL
30 mL
60 mL
Product Type:
Luer Lock
Luer Slip
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Product Type
14817178 10 mL Luer Lock
14817179 1 mL Luer Slip
14817109 3 mL Luer Lock
14817180 5 mL Luer Slip
14817181 10 mL Luer Slip
14817110 5 mL Luer Lock
14817111 20 mL Luer Lock
14817210 3 mL Luer Slip
14817112 30 mL Luer Lock
14817113 60 mL Luer Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14817178 Supplier Air-Tite™ Supplier No. ML10

Air-Tite™ Sterile Syringes are high quality, yet economical for laboratory and veterinary use only

  • Smooth plunger
  • Bold graduations
  • Disposable
  • Sterile
  • Polypropylene
  • Economical
  • Single-use
  • Latex Free
  • ISO Compatible

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ISO 13485
Material Polyisoprene Rubber (Tip), Polypropylene, Polypropylene (Barrel), Polypropylene (Plunger)
Color Clear
For Use With (Application) Laboratory / Veterinary
Latex Free Yes
Product Type Luer Lock
Disposable Single-use
Packaging Individually Wrapped
Volume (Metric) 10 mL
Connection Type Luer Lock
Graduated Yes
Sterility Sterile
Show More Show Less

Not for human use - laboratory and veterinary use only