Learn More
Contec™ Sterile Polynit Heatseal LE Wipes
GREENER_CHOICE
Polynit Heatseal Low Endotoxin Wipes are manufactured from high quality 100% knitted polyester fabric with sealed edges and produce low levels of particles and fibers.
$751.42 - $793.50
Specifications
Product Type Wipe
Material Polyester Knit
Basis Weight 140
Sterility Sterile
Grade A/B
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19033865
GREENER_CHOICE
View Documents Promotion Details
Contec™
LWLE0001
9 x 9 in.
Case of 500 Each for $793.50
Only null left
19200056
GREENER_CHOICE
View Documents Promotion Details
Contec™
LWLE0002
12 x 12 in.
Case of 360 Each for $751.42
Only null left
Description
Description
  • Polyester fabric offers a wide range of solvent compatibility
  • Validated sterile to 10-6 Sterility Assurance Level
  • Ideal for use in product contact areas
  • Half folded and triple packaged in linear tear outer bags for ease of transfer into ISO Class 5 (Grades A/B) cleanrooms.
  • Small packaging takes up less space making it ideal for any side cleanroom.
  • Recommended for Grade A/B or ISO Class 5 environments - product contact areas
  • 100% laundered knitted polyester generates very low levels of particles and fibers
  • Laser sealed edges reduces fiber generation from the edge of the wipe
  • Certified endotoxin level of <1 EU per wipe making them suitable for use in product contact areas
  • Smaller number of wipes per bag eliminates waste
Specifications
Specifications
Wipe
140
A/B
Polyester Knit
Sterile