missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab Sterile Phosphate Tuberculosis Buffer Packets
Description
- Sterile
- Phosphate in a dry powder form
- 5 packets/package
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffers |
| For Use With (Application) | Sputum Testing |
| Physical Form | Solution |
| Quantity | 5/Pk. |
| Formulation | Dry Powder |
| Packaging | 4.8 g Packets |
| Sterility | Sterile |