missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Sterile PD-Tip™ II Precision Dispenser Tips for Repeating Pipettes

BRAND™ Sterile PD-Tip™ II Precision Dispenser fits all instruments using standard repeating tips.

Supplier:  BRANDTECH™ BRAND™ 705734

 View more versions of this product

Catalog No. 14380989


Only null left

Description

Description

  • Automatic size recognition by BRAND™ HandyStep™ S, HandyStep™ Touch S and HandyStep™ Electronic pipettes
  • BIO-CERT™ sterile
  • BIO-CERT™ tips are free of DNA, RNase, endotoxin and ATP
  • 25 mL and 50 mL sizes include one adapter
  • Adapters are autoclavable at 121°C for 20 minutes
These tips are suitable for the following multiple dispensers:
  • BRAND Handystep™ S, HandyStep™ electronic, HandyStep™ touch/ S
  • Eppendorf Multipette™ 4780
  • Gilson Distriman™ and Gilson Repetman™
  • Ritter Ripette™ pro
  • EDOS™ 5221
  • Rainin AutoRep™ E, Rainin AutoRep™ S, Rainin AutoRep™ M
  • STARLAB RepeatOne™
Specifications

Specifications

1 mL
Sterile
Not Autoclavable
Free from endotoxin, DNA, Rnase, ATP
Individually Wrapped
100/Pk
Precision Dispenser Tips
HandyStep Touch, HandyStep Touch S
PD-Tip II
Non-filtered
Natural
Certificate of Performance/Purity Certificate
Beta
Positive Displacement
100
Single-use
Yes
Polypropylene, Polyethylene (Filter)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions