missing translation for 'onlineSavingsMsg'
Learn More

Air-Tite™ Sterile Hypodermic Needles

Catalog No. 14817203
Change view
Click to view available options
Needle Gauge:
14 G
16 G
18 G
20 G
21 G
22 G
23 G
25 G
26 G
27 G
Length (English) Needle:
0.25 in.
0.50 in.
0.625 in.
0.75 in.
1 in.
1.5 in.
2 in.
3.25 in.
3.5 in.
4 in.
Certifications/Compliance:
ISO 13485
ISO 9001 Certified
Color:
Black
Blue
Brown
Gray
Green
Orange
Pink
White
Yellow
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (English) Needle Certifications/Compliance Color
14817203 14 G 3.25 in. ISO 13485 Blue
14817144 14 G 1.5 in. ISO 9001 Certified Blue
14817164 22 G 3.5 in. ISO 9001 Certified Black
14817208 20 G 1 in. ISO 13485 Yellow
14817145 14 G 2 in. ISO 9001 Certified Blue
14817209 20 G 1.5 in. ISO 13485 Yellow
14817146 16 G 0.75 in. ISO 9001 Certified White
14817204 16 G 4 in. ISO 13485 White
14817147 16 G 1 in. ISO 9001 Certified White
14817148 18 G 0.25 in. ISO 9001 Certified Pink
14817205 18 G 1.5 in. ISO 13485 Pink
14817206 18 G 2 in. ISO 13485 Pink
14817151 18 G 1 in. ISO 9001 Certified Pink
14817159 21 G 1.5 in. ISO 9001 Certified Green
14817161 22 G 0.75 in. ISO 9001 Certified Black
14817162 22 G 1 in. ISO 9001 Certified Black
14817163 22 G 1.5 in. ISO 9001 Certified Black
14817166 23 G 1 in. ISO 9001 Certified Blue
14817167 25 G 0.625 in. ISO 9001 Certified Orange
14817168 25 G 1 in. ISO 9001 Certified Orange
14817169 25 G 1.5 in. ISO 9001 Certified Orange
14817170 26 G 0.50 in. ISO 9001 Certified Brown
14817171 27 G 0.50 in. ISO 9001 Certified Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. 14817203 Supplier Air-Tite™ Supplier No. AIRN14314
Only null left

Air-Tite™ Sterile Hypodermic Needles are high quality, yet economical for laboratory and veterinary use only

  • Disposable Needles
  • Sterile
  • Polypropylene Hub
  • Stainless Steel Cannula
  • Single-use
  • Latex Free
  • ISO
  • Lab/Vet

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ISO 13485
Material Polypropylene (Hub), Stainless Steel (Cannula)
Color Blue
Needle Gauge 14 G
For Use With (Application) Laboratory / Veterinary
Latex Free Yes
Length (English) Needle 3.25 in.
Disposable Single-use
Packaging Individually Wrapped
Needle Point Style Tri-Bevel
Connection Type Luer Lock
Graduated No
Sterility Sterile
Show More Show Less

Not for human use - laboratory and veterinary use only