Learn More
Sklar Instruments Econo Sterile Iris Scissors, Sklar , 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, Disposable. CS/25.
Catalog No. NC0477393
Encompass
Econo Sterile Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, Disposable. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. BX25.
Catalog No. NC0477393 Supplier Sklar Instruments Supplier No. 962507
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Econo Sterile Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, Disposable. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. BX25.