Learn More
Greiner Bio-One Sterile Cotton Swabs
Catalog No. 07000547
Change view
Click to view available options
Quantity:
1400/Cs.
Catalog No. Quantity
07000547 1400/Cs.
1 options
Catalog No. 07000547 Supplier Greiner Bio-One Supplier No. 421084
Only null left
Sterilized by irradiation. Greiner Bio-One Sterile Cotton Swab is suitable for collection and transport of non-human bacteriological, serological and cytological samples.
  • Sterilised by irradiation
  • Height: 145mm (5.7 in.)
  • For hygienic controls in food industry and environmental sampling
Specifications
Description Individually packaged
Sterility Sterile
Product Type Swab
Material Cotton
Quantity 1400/Cs.
For Use With (Application) Collection and transport of non-human bacteriological, serological and cytological samples
For research use only. Not for use in diagnostic procedures, unless otherwise indicated.