Learn More
Biospec Products Inc STERILE, 2ML VIAL, 500/PK
Supplier: BioSpec Products Inc 522S
Our 2.0 ml, sterile screw cap with o-ring seal, microcentrifuge vials are made of inert polypropylene and have straight walls with a sharp conical bottom.