Learn More
Fisherbrand™ Round-Bottom Polystyrene Test Tubes With Cap
Catalog No. FB1496120
Change view
Click to view available options
Capacity (Metric):
5 mL
8 mL
14 mL
16 mL
19 mL
Sterility:
Non-sterile
Sterile
Closure Type:
Screw Cap
Snap Cap
Packaging:
Bag (8 Pk. of 125 Tubes Ea.)
Blister Pk.
Blister Pk. (20 Pk. of 25 Tubes Ea.)
Blister Pk. (8 Pk. of 125 Tubes Ea.)
Bulk
Individually wrapped
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Closure Type Packaging
FB1496120 5 mL Non-sterile Snap Cap Bulk
FB149563B 5 mL Sterile Snap Cap Individually wrapped
FB149563D 5 mL Sterile Snap Cap Bag (8 Pk. of 125 Tubes Ea.)
FB149563C 5 mL Sterile Snap Cap Blister Pk. (20 Pk. of 25 Tubes Ea.)
FB149564A 8 mL Sterile Screw Cap Blister Pk.
FB149566B 14 mL Sterile Snap Cap Blister Pk. (8 Pk. of 125 Tubes Ea.)
FB149566D 14 mL Sterile Snap Cap Blister Pk. (20 Pk. of 25 Tubes Ea.)
FB149566A 14 mL Sterile Snap Cap Individually wrapped
FB149565A 16 mL Sterile Screw Cap Individually wrapped
FB149565B 16 mL Sterile Screw Cap Blister Pk.
FB149565C 19 mL Sterile Screw Cap Individually wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. FB1496120
Get Up to $2,500 in Products with Purchase
View all promotions
Fisherbrand™ round-bottom tubes provide reliable containment for various laboratory fluid samples and preparations.
  • Ideal for use in laboratory protocols requiring round bottom tubes
  • Ensures reliable handling of liquid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability, while polystyrene tubes offer high optical clarity
  • Available with and without snap cap - dual-position snap caps offer both vented and fully closed positions.
  • 3,000 RCF rating for polypropylene tubes, 1,400 RCF rating for polystyrene tubes
  • Dimensions and volumes are approximate
Specifications
Color Clear
Capacity (Metric) 5 mL
Length (Metric) 75 mm
Material Polystyrene
Disposable Single-use
Packaging Bulk
Sterility Non-sterile
Product Line Fisherbrand
Graduated No
Closure Type Snap Cap
Product Type Test Tube
Show More Show Less