Learn More
Fisherbrand™ Round-Bottom Polystyrene Test Tubes With Cap
Fisherbrand™ round-bottom tubes provide reliable containment for various laboratory fluid samples and preparations.
 View more versions of this product
Catalog No. FB149566B
Only null left
Description
Description
  • Ideal for use in laboratory protocols requiring round bottom tubes
  • Ensures reliable handling of liquid samples
  • Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability, while polystyrene tubes offer high optical clarity
  • Available with and without snap cap - dual-position snap caps offer both vented and fully closed positions.
  • 3,000 RCF rating for polypropylene tubes, 1,400 RCF rating for polystyrene tubes
  • Dimensions and volumes are approximate
  • Graduated
Specifications
Chemical Identifiers
Clear
Single-use
Test Tube
Polystyrene
Fisherbrand
Specifications
Clear
100 mm
Single-use
Sterile
Yes
Test Tube
14 mL
Polystyrene
Blister Pk. (8 Pk. of 125 Tubes Ea.)
Fisherbrand
Snap Cap
Product Suggestions
Product Suggestions