missing translation for 'onlineSavingsMsg'
Learn More

VAI STERI-BUFFER™ Phosphate Buffer
GREENER_CHOICE

Catalog No. 19546613
Change view
Click to view available options
STERI-BUFFER™ Phosphate Buffer; Quantity: 90 mL (3 oz.); Sterile: Yes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19546613 STERI-BUFFER™ Phosphate Buffer; Quantity: 90 mL (3 oz.); Sterile: Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19546613 Supplier VAI Supplier No. SB10090

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Certificate of Analysis and Certificate of Sterility

STERI-BUFFER™ Phosphate Buffer for disinfectant preparation and dilution.

  • Buffered to a pH of 7.2 ± 0.2
  • Made with USP water for injection filtered at 0.2 μm
  • Filled in easy open-and-close wide-mouth bottles that are scaled on four sides for easy measurement
  • Terminally sterilized through a validated cycle at a SAL level of 10-6
  • Sealed with a no-tamper strip that is broken once the bottle is opened
  • Filled in a ISO 5 cleanroom
  • Validated for sterility

Order Info

Shelf life of two years. Not for parenteral administration.

Compliance

Lot sterility tested per current USP compendium.

Specifications

Chemical Name or Material Phosphate Buffer
For Use With (Application) For disinfectant preparation and dilution
pH 7.2 ± 0.2
Quantity 90 mL (3 oz.)
Sterility Sterile
Includes Certificate of Analysis, certificate of Sterility