missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™ MSC SFM XenoFree

Catalog No. A1067501
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1067501 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1067501 Supplier Gibco™ Supplier No. A1067501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

StemPro™ MSC SFM XenoFree

GIBCO™ StemPro™ MSC SFM XenoFree has been developed for the growth and expansion of human mesenchymal stem cells (MSCs) and Adipose-derived Stem Cells (ADSCs) under completely serum-free and xenogeneic (xeno)-free conditions. Using StemPro™ MSC SFM XenoFree, human MSCs or ADSCs can be expanded for multiple passages while maintaining their multipotent phenotype (i.e. ability to differentiate into osteogenic, chondrogenic and adipogenic lineages). StemPro™ MSC SFM XenoFree is cGMP compliant which allows for traceability and manufacturing reliability.

Specifications

Cell Type Mesenchymal Stem Cells
Classification Xeno-free
Form Liquid
Product Type Stem Cell Serum Free Media (SFM)
Serum Level Serum-free
Without Additives No Glutamine, No Phenol Red
Product Line StemPro
Quantity 1 Kit
Included Antibiotics No antibiotics
Purity or Quality Grade MS Cell-Qualified
Species Human
Content And Storage Each kit contains 1 x 500 ml StemPro™ MSC SFM Basal Medium and 1 X 5 ml StemPro™ MSC SFM XenoFree Supplement. StemPro™ MSC SFM Basal Medium is stored at 2 to 8°C and stable for 12 months. The StemPro™ MSC SFM XenoFree Supplement is stored frozen at -5 to -20°C
Culture Environment CO2
Culture Type Adherent Cell Culture
Show More Show Less
What is the concentration of sodium bicarbonate in StemPro MSC SFM XenoFree?

StemPro MSC SFM XenoFree contains 2.2 g/L of sodium bicarbonate.

For Research Use Only. Not for use in diagnostic procedures.