Learn More
Stemcell Technologies Inc STEMdiff™ Hematopoietic - EB Supplement B, Size: 7 mL
Catalog No. NC2043818
Encompass
STEMdiff™ T Cell Kit includes serum-free media and supplements for the feeder-free differentiation of human pluripotent stem cells (hPSCs) to T cells.STEMdiff™ Hematopoietic - EB reagents are animal component-free and optimized for lymphoid differentiation potential. Using these reagents, CD34 hematopoietic progenitor cells are first generated from hPSCs; these can then be differentiated to T cells using the StemSpan™ components.
Catalog No. NC2043818 Supplier Stemcell Technologies Inc Supplier No. 1000173
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
STEMdiff™ T Cell Kit includes serum-free media and supplements for the feeder-free differentiation of human pluripotent stem cells (hPSCs) to T cells.STEMdiff™ Hematopoietic - EB reagents are animal component-free and optimized for lymphoid differentiation potential. Using these reagents, CD34 hematopoietic progenitor cells are first generated from hPSCs; these can then be differentiated to T cells using the StemSpan™ components.