Learn More
Stemcell Technologies STEMdiff™ APEL™2 Medium, Size: 100 mL DFS Item
Supplier:  Stemcell Technologies Inc 05270
STEMdiff™ APEL™ 2 Medium is a fully defined, serum-free and animal component-free medium for the differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. It is based on the APEL formulation published by Dr. Andrew Elefanty and lacks undefined components such as protein-free hybridoma medium.STEMdiff™ APEL™ 2 can be used in adherent or embryoid body (EB)-based protocols, such as with AggreWell™. It can be used with a variety of different induction factors or cytokines to support differentiation along ectoderm, mesoderm and endoderm lineages.
Catalog No. NC1160175
Only null left
Explore available promotions