Learn More
Fine Science Tools STEEL BURRS 1.4MM
Catalog No. NC9727231
Encompass
Tip Diameter: 1.4mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Carbon Steel/ Length: 44mm/ Autoclave Safe: No
Catalog No. NC9727231 Supplier Fine Science Tools Supplier No. 1900714
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tip Diameter: 1.4mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Carbon Steel/ Length: 44mm/ Autoclave Safe: No