missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAI Steel-Bright Stainless-Steel Cleaner
Description
- For removal of chemical residues, spotting and staining on stainless-steel surfaces without leaving a powdery residue.
- Ideal on stainless steel, chromes, brass aluminum, or copper.
- Filtered at 0.2 μ
- Gamma irradiated
Order Info
Double-bagged packaging. Delivered with lot specific Certificate of Analysis.
Specifications
Specifications
| Sterility | Sterile |
| Dimensions (L x W) | 12 x 12 in. |
| Format | Wipes |
| For Use With (Application) | For aseptic manufacturing, removes chemical residues, spotting and staining |