missing translation for 'onlineSavingsMsg'
Learn More

VAI Steel-Bright Stainless-Steel Cleaner

Catalog No. 19547079
Change view
Click to view available options
Format:
Aerosol Spray
Wipes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19547079 Aerosol Spray
19547081 Wipes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19547079 Supplier VAI Supplier No. SB02
Only null left

Steel-Bright Stainless-Steel Cleaner is an emulsion-based cleaner that will not rainbow or accumulate a heavy build up.

  • For removal of chemical residues, spotting and staining on stainless-steel surfaces without leaving a powdery residue.
  • Ideal on stainless steel, chromes, brass aluminum, or copper.
  • Filtered at 0.2 μ
  • Gamma irradiated

Order Info

Double-bagged packaging. Delivered with lot specific Certificate of Analysis.

Specifications

Format Aerosol Spray
For Use With (Application) For aseptic manufacturing, removes chemical residues, spotting and staining
Volume (English) 8 oz.
Volume (Metric) 236.6 mL