Learn More
Supertek™ Steel Ball, Drilled
GREENER_CHOICE
These drilled balls are excellent for use as pendulum bobs or in impact and collision experiments.
$8.11 - $13.00
Specifications
Product Type Steel Ball
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Price Quantity & Availability  
S012335
GREENER_CHOICE
Encompass
View Documents Promotion Details
Supertek™
PH10594D/3SP
0.75 in.
Each for $8.11
Only null left
S012334
GREENER_CHOICE
Encompass
View Documents Promotion Details
Supertek™
PH10594D/5SP
1 in.
Each for $13.00
Only null left
Description
Description
  • Drilled with 3 mm hole.
  • Pendulum bobs.
  • Use in impact and collision experiments.
Specifications
Specifications
Steel Ball