missing translation for 'onlineSavingsMsg'
Learn More

Stearylamine 85.0+%, TCI America™

Catalog No. O001425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O001425G 25 g
O0014500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O001425G Supplier TCI America Supplier No. O001425G

Chemical Identifiers

CAS 124-30-1
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.52
MDL Number MFCD00008159
InChI Key REYJJPSVUYRZGE-UHFFFAOYSA-N
Synonym octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm
PubChem CID 15793
ChEBI CHEBI:63866
IUPAC Name octadecan-1-amine
SMILES CCCCCCCCCCCCCCCCCCN

Specifications

Melting Point 53°C
Color White
Boiling Point 349°C
Quantity 25 g
UN Number 3077
Formula Weight 269.52
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Stearylamine
EINECSNumber (2)-0133&(2)-0176&(2)-0185
RTECSNumber RG4150000
TSCA Yes