missing translation for 'onlineSavingsMsg'
Learn More

Stearyl stearate, 98%

Catalog No. p-2692228
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC368110010 1 g Glass bottle
AC368110050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC368110010 Supplier Thermo Scientific Chemicals Supplier No. 368110010
Only null left

CAS: 2778-96-3 | C36H72O2 | 536.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2778-96-3
Molecular Formula C36H72O2
Molecular Weight (g/mol) 536.97
MDL Number MFCD00056225
InChI Key NKBWPOSQERPBFI-UHFFFAOYSA-N
Synonym stearyl stearate, octadecyl stearate, octadecanoic acid, octadecyl ester, stearic acid stearyl ester, unii-5wx2egd0dk, stearic acid, stearyl ester, octadecanyl octadecanoate, 5wx2egd0dk, stearic acid, octadecyl ester, cyclochem ss
PubChem CID 17720
IUPAC Name octadecyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)16COO(CH2)17CH3
Quantity 1 g
Formula Weight 536.97
Percent Purity 98%
Physical Form Powder or Platelets
Chemical Name or Material Stearyl stearate, 98%
EINECSNumber 220-476-5

RUO – Research Use Only