missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stearyl Methacrylate (stabilized with MEHQ) 97.0+%, TCI America™
$48.46 - $134.21
Chemical Identifiers
| CAS | 32360-05-7 |
|---|---|
| Molecular Formula | C22H42O2 |
| Molecular Weight (g/mol) | 338.576 |
| MDL Number | MFCD00026683 |
| InChI Key | HMZGPNHSPWNGEP-UHFFFAOYSA-N |
| Synonym | Methacrylic Acid Octadecyl Ester, Octadecyl Methacrylate, Methacrylic Acid Stearyl Ester |
| PubChem CID | 122600 |
| IUPAC Name | octadecyl 2-methylprop-2-enoate |
| SMILES | CCCCCCCCCCCCCCCCCCOC(=O)C(=C)C |
Chemical Identifiers
| 32360-05-7 | |
| 338.576 | |
| HMZGPNHSPWNGEP-UHFFFAOYSA-N | |
| 122600 | |
| CCCCCCCCCCCCCCCCCCOC(=O)C(=C)C |
| C22H42O2 | |
| MFCD00026683 | |
| Methacrylic Acid Octadecyl Ester, Octadecyl Methacrylate, Methacrylic Acid Stearyl Ester | |
| octadecyl 2-methylprop-2-enoate |
Specifications
| 32360-05-7 | |
| 235°C | |
| MFCD00026683 | |
| Methacrylic Acid Octadecyl Ester, Octadecyl Methacrylate, Methacrylic Acid Stearyl Ester | |
| CCCCCCCCCCCCCCCCCCOC(=O)C(=C)C | |
| 338.576 | |
| 338.58 | |
| Liquid |
| Yellow | |
| C22H42O2 | |
| 25 g | |
| HMZGPNHSPWNGEP-UHFFFAOYSA-N | |
| octadecyl 2-methylprop-2-enoate | |
| 122600 | |
| ≥97.0% (GC) | |
| Stearyl Methacrylate (stabilized with MEHQ) |
Safety and Handling
EINECSNumber : (2)-1039
TSCA : Yes