missing translation for 'onlineSavingsMsg'
Learn More

Stearyl Methacrylate (stabilized with MEHQ) 97.0+%, TCI America™

Catalog No. M0593400G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0593400G 400 g
M059325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0593400G Supplier TCI America Supplier No. M0593400G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32360-05-7
Molecular Formula C22H42O2
Molecular Weight (g/mol) 338.576
MDL Number MFCD00026683
InChI Key HMZGPNHSPWNGEP-UHFFFAOYSA-N
Synonym Methacrylic Acid Octadecyl Ester, Octadecyl Methacrylate, Methacrylic Acid Stearyl Ester
PubChem CID 122600
IUPAC Name octadecyl 2-methylprop-2-enoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)C(=C)C

Specifications

Color Yellow
Boiling Point 235°C
Quantity 400 g
Formula Weight 338.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Stearyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1039
TSCA Yes