missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stearyl Glycyrrhetinate 98.0+%, TCI America™
Supplier: TCI America S098825G
Specifications
| Stearyl Glycyrrhetinate | |
| 77°C | |
| C48H82O4 | |
| 25 g | |
| WNIFXKPDILJURQ-OVDQTPITSA-N | |
| octadecyl (2S,4aS,6aR,6bR,10S,12aS)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate | |
| 122130446 | |
| ≥98.0% (HPLC) |
| 13832-70-7 | |
| White-Yellow | |
| MFCD00674112 | |
| Glycyrrhetinic Acid Stearyl Ester, Octadecyl Glycyrrhetinate, Glycyrrhetinic Acid Octadecyl Ester | |
| CCCCCCCCCCCCCCCCCCOC(=O)C1(CCC2(CCC3(C(=CC(=O)C4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C)C | |
| 723.18 | |
| 723.18 | |
| Crystalline Powder |
Chemical Identifiers
| 13832-70-7 | |
| 723.18 | |
| WNIFXKPDILJURQ-OVDQTPITSA-N | |
| 122130446 | |
| CCCCCCCCCCCCCCCCCCOC(=O)C1(CCC2(CCC3(C(=CC(=O)C4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1)C)C)C |
| C48H82O4 | |
| MFCD00674112 | |
| Glycyrrhetinic Acid Stearyl Ester, Octadecyl Glycyrrhetinate, Glycyrrhetinic Acid Octadecyl Ester | |
| octadecyl (2S,4aS,6aR,6bR,10S,12aS)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate |
Safety and Handling
EINECSNumber : (9)-0722
TSCA : No