missing translation for 'onlineSavingsMsg'
Learn More

Stearyl Gallate 97.0+%, TCI America™

Catalog No. g001925g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G001925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G001925G Supplier TCI America Supplier No. G001925G
Only null left

Chemical Identifiers

CAS 10361-12-3
Molecular Formula C25H42O5
Molecular Weight (g/mol) 422.61
MDL Number MFCD00016429
InChI Key BRNPAEUKZMBRLQ-UHFFFAOYSA-N
Synonym Gallic Acid Stearyl Ester
PubChem CID 473591
IUPAC Name octadecyl 3,4,5-trihydroxybenzoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)C1=CC(O)=C(O)C(O)=C1

Specifications

Melting Point 104°C
Quantity 25 g
Formula Weight 422.61
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Stearyl Gallate
EINECSNumber (3)-1584
TSCA No