missing translation for 'onlineSavingsMsg'
Learn More

Stearyl Acrylate (stabilized with MEHQ) 97.0+%, TCI America™

Catalog No. A101125G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A101125G 25 g
A1011400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A101125G Supplier TCI America Supplier No. A101125G
Only null left

Chemical Identifiers

CAS 4813-57-4
Molecular Formula C21H40O2
Molecular Weight (g/mol) 324.55
MDL Number MFCD00015091
InChI Key FSAJWMJJORKPKS-UHFFFAOYSA-N
Synonym Acrylic Acid Octadecyl Ester, Acrylic Acid Stearyl Ester, Octadecyl Acrylate
PubChem CID 62552
IUPAC Name octadecyl prop-2-enoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)C=C

Specifications

Melting Point 28°C
Color White
Quantity 25 g
Formula Weight 324.55
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Stearyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0990
TSCA Yes