missing translation for 'onlineSavingsMsg'
Learn More

Stearyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate 98.0+%, TCI America™

Catalog No. D1644500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1644500G 500 g
D164425G 25 g
D1644100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1644500G Supplier TCI America Supplier No. D1644500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2082-79-3
Molecular Formula C35H62O3
Molecular Weight (g/mol) 530.878
MDL Number MFCD00017498
InChI Key SSDSCDGVMJFTEQ-UHFFFAOYSA-N
Synonym 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid Stearyl Ester, Octadecyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate
PubChem CID 16386
IUPAC Name octadecyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

Melting Point 53°C
Color White
Quantity 500 g
Formula Weight 530.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Stearyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate
EINECSNumber (3)-1737
TSCA Yes