missing translation for 'onlineSavingsMsg'
Learn More

Stearoyl Chloride 97.0+%, TCI America™

Catalog No. p-7135882
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S040425G 25 g
S0404250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S040425G Supplier TCI America Supplier No. S040425G
Only null left

Chemical Identifiers

CAS 112-76-5
Molecular Formula C18H35ClO
Molecular Weight (g/mol) 302.927
MDL Number MFCD00000744
InChI Key WTBAHSZERDXKKZ-UHFFFAOYSA-N
PubChem CID 8212
IUPAC Name octadecanoyl chloride
SMILES CCCCCCCCCCCCCCCCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 215°C
Quantity 25 g
UN Number 3265
Formula Weight 302.93
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Stearoyl Chloride
EINECSNumber (2)-0632&(2)-3552
RTECSNumber RG2162000
TSCA Yes