missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stearic Anhydride 97.0+%, TCI America™
$195.73 - $1541.57
Chemical Identifiers
| CAS | 638-08-4 |
|---|---|
| Molecular Formula | C36H70O3 |
| Molecular Weight (g/mol) | 550.953 |
| MDL Number | MFCD00009004 |
| InChI Key | WVJVHUWVQNLPCR-UHFFFAOYSA-N |
| Synonym | stearic anhydride, octadecanoic acid, anhydride, stearic acid anhydride, stearicanhydride, octadecanoic acid, 1,1'-anhydride, octadecanoic anhydride, e r degrees eeaou, octadecanoic anhydride #, acmc-1b4gn, octadecanoic acid,1,1'-anhydride |
| PubChem CID | 69485 |
| IUPAC Name | octadecanoyl octadecanoate |
| SMILES | CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC |
Chemical Identifiers
| 638-08-4 | |
| 550.953 | |
| WVJVHUWVQNLPCR-UHFFFAOYSA-N | |
| 69485 | |
| CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC |
| C36H70O3 | |
| MFCD00009004 | |
| stearic anhydride, octadecanoic acid, anhydride, stearic acid anhydride, stearicanhydride, octadecanoic acid, 1,1'-anhydride, octadecanoic anhydride, e r degrees eeaou, octadecanoic anhydride #, acmc-1b4gn, octadecanoic acid,1,1'-anhydride | |
| octadecanoyl octadecanoate |
Specifications
| 638-08-4 | |
| White | |
| MFCD00009004 | |
| stearic anhydride, octadecanoic acid, anhydride, stearic acid anhydride, stearicanhydride, octadecanoic acid, 1,1'-anhydride, octadecanoic anhydride, e r degrees eeaou, octadecanoic anhydride #, acmc-1b4gn, octadecanoic acid,1,1'-anhydride | |
| CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC | |
| 550.953 | |
| 550.95 | |
| Crystalline Powder |
| 71°C | |
| C36H70O3 | |
| 25 g | |
| WVJVHUWVQNLPCR-UHFFFAOYSA-N | |
| octadecanoyl octadecanoate | |
| 69485 | |
| ≥97.0% (T) | |
| Stearic Anhydride |
Safety and Handling
TSCA : Yes