missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Steam High Performance Pipe Marker, 8 to 10 in., Black on Yellow
Description
- Designed for use on rusty, dirty, wet or rough pipes where pressure sensitive adhesive labels aren’t effective.
- Resistant to UV light exposure, high humidity, high temperatures, abrasion, chemical reagents and physical agitation.
- Excellent chemical resistance.
- Durable corner grommets resist tears and rips to keep marker firmly in place using nylon ties.
- Comes with two large nylon ties.
Specifications
Specifications
| Adhesive Type | Non-adhesive |
| Coating | Laminate |
| Color | Black on Yellow |
| Display | Text |
| Finish | Gloss |
| Format | Sheet |
| Material | Coiled Plastic (B-689) |
| Product Type | Pipe Marker |
| Sterility | Non-sterile |
| Dimensions (L x W) | 10 x 24 in. |
| Show More |