Learn More
Eisco™ Steam Engine Model
Catalog No. S23747
Change view
Click to view available options
14.5 x 7 x 4 in.
Catalog No.
S23747 14.5 x 7 x 4 in.
1 options
Catalog No. S23747 Supplier Eisco™ Supplier No. PH0483
Shows the internal structure and operating principles of a working steam engine.
  • Complete with working parts of piston, slide valve, link motion, reversing
  • Cross-section cut metal cylinder
  • Covered with clear plate to show movement of piston inside cylinder
  • Movement of wheel light enough to demonstrate even by blowing air from mouth
  • Mounted on base
Specifications
Product Type Model, Steam Engine
Description Cross-section cut metal cylinder
For Use With (Application) Shows the internal structure and operating principles of a working steam engine.
Includes working parts of piston, slide valve, link motion, reversing
Dimensions (L x W) 14.5 x 7 x 4 in.