missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Stealth RNAi™ siRNA Negative Control Kit

Catalog No. 12935100
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12935100 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12935100 Supplier Invitrogen™ Supplier No. 12935100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stealth RNAi™ siRNA Negative Control Kit

Stealth RNAi™ siRNA Negative Controls are a great way to measure the effect of your experimental Stealth RNAi™ siRNA duplex versus background. These controls are:

  • Designed in 3 levels of GC content (Lo, Med, Hi) with 3 sequences available per level so researchers can match GC content to their experimental Stealth RNAi™ siRNA
  • Not homologous to anything in the vertebrate transcriptome
  • Tested not to induce stress response Stealth RNAi™ siRNA

    Negative Control Kit contains all three controls (Lo, Med, Hi GC content-excluding sequences #2 and #3) and each is also available separately. Get the ideal negative controls for your RNAi experiment so you can assess your results accurately.
  • Specifications

    Content And Storage Each individual control is supplied as 250 μl at 20 μM. The Stealth RNAi™ siRNA Negative Control Kit contains one each of all three negative controls (each 250 μl @ 20 μM). Store at -20°C. Products are guaranteed stable for 6 months when properly stored.
    Control Type Negative Control
    RNAi Type siRNA
    For Use With (Application) RNAi, Transfection
    Label or Dye Unconjugated
    Product Line Stealth RNAi™ siRNA
    Product Type siRNA
    Quantity 1 Kit
    What are the benefits of using a vector to deliver RNAi?

    Vector technologies allow you to:

    Achieve transient or stable target knockdown
    Perform RNAi in any cell type, even hard-to-transfect, primary, and non-dividing cells
    Regulate gene inhibition with inducible siRNA expression
    Select for a pure population of cells stably expressing an siRNA sequence
    Control gene expression in vivo with tissue-specific promoters

    Why are my cells dying after transfection?

    We would suggest running a transfection reagent control only to determine if your cells are sensitive to the transfection reagent. Additionally, you can try using different cell densities and siRNA concentrations to diminish any toxic effects from the transfection itself.

    I transfected my siRNA and the mRNA levels are down, but the protein is not. Why is that?

    In some cases, knockdown of a protein can be affected by other variables such as protein turnover rate, even though the RNA is knocked down. Additionally, a longer time course may be needed to see an effect on protein compared to mRNA.

    I am not getting my target knockdown. What could be the cause of this?

    Please see the following possibilities and suggestions:

    - How many siRNA did you test? Is there any knockdown? If there is no knockdown (<10%) in any of the siRNA, then the assay is likely the problem. Try using a different qRT-PCR assay to assess knockdown.
    - What was the positioning of the qRT-PCR assay target site relative to the cut site for the siRNA? If greater than 3,000 bases away, the problem could be alternative splice transcripts.
    - What are the Cts for the experiment? They should be below 35 in a 40-cycle qRT-PCR experiment.
    - Did you confirm the siRNA got into the cell? We recommend using a validated positive control siRNA to check the transfection efficiency.

    I am not getting any knockdown with my siRNA. What do you suggest I try?

    Please see the following possibilities and suggestions:

    - Were the mRNA levels checked? The most reliable method is real-time PCR. In some cases, knockdown of a protein can be affected by other variables, such as protein turnover rate, even though the RNA is knocked down.
    - How is the RNA being isolated? Has the quality of the isolated RNA been checked? Ensure that the RNA has not been degraded.
    - Was a positive control used? This can help to determine whether the reagents are working and whether the siRNA was delivered correctly to the cell. Run your experiment in parallel with the positive control siRNA.
    - Was a transfection control used? What is the percentage of transfected cells?
    - Was a time course used? Generally, gene silencing can be assessed as early as 24 hours posttransfection. However, the duration and level of knockdown are dependent on cell type and concentration of siRNA.
    - Was optimization of transfection conditions performed? You can try using different cell densities and siRNA concentrations.
    - Which concentration of siRNA did you use? We recommend testing multiple concentrations between 5 nM and 100 nM.

    When do I evaluate knockdown after transfection of siRNA?

    Depending on the gene you are working with, it can be measured at the mRNA level as soon as a few hours after transfection. We recommend assessing the mRNA knockdown 48 hours posttransfection. Factors affect the timing include the transcription activity, the turnover rate for the mRNA, and if there are alternative pathways. To determine the peak knockdown, it is best to perform a time course experiment.

    What should I dissolve my RNA pellet in, water or buffer?

    We recommend dissolving the single stranded RNA in 1X TE buffer (prepared under RNase-free conditions (10 mM TrisCl, pH 7.5, 0.1 mM EDTA). This buffers the pH and chelates metal ions that can contribute to RNA degradation. RNase-free water is also acceptable. Duplex RNA (siRNA) comes lyophilized from 10 mM Tris-HCl, pH 8.0, 20 mM NaCl, 1 mM EDTA. Resuspending in the appropriate amount of nuclease free water to bring the RNA conc. to 20 µM will reconstitute the buffer to the same.

    How stable are the siRNAs/miRNAs?

    As a dry pellet they can be stored at -20 degrees C for 6 months.

    What is the molecular weight of Stealth RNAi and BLOCK-iT Fluorescent Oligo?

    The average molecular weight is 16,100 g/mol.

    What are the positive and negative controls I should use with my Stealth RNAi siRNA experiment?

    We offer several negative and positive controls for Stealth siRNA:

    For Stealth Positive Control:
    - Stealth RNAi siRNA GAPDH Positive Control (human) (Cat. No. 12935140, 250 µL)
    - Stealth RNAi siRNA beta-Lactamase Reporter Control (Cat. No. 12935148, 250 µL)
    - Stealth RNAi siRNA GFP Reporter Control (Cat. No. 12935145, 250 µL)
    - Stealth RNAi siRNA Luciferase Reporter Control (Cat. No. 12935146, 250 µL)

    For Stealth Negative Control:
    Hi (55-65% GC), Med (45-55% GC), or Low (35-45% GC) Duplexes are offered:

    - Stealth RNAi siRNA Negative Control Hi GC (Cat. No. 12935400, 250 µL)
    - Stealth RNAi siRNA Negative Control Hi GC Duplex #2 (Cat. No. 12935114, 250 µL)
    - Stealth RNAi siRNA Negative Control Hi GC Duplex #3 (Cat. No. 12935115, 250 µL)
    - Stealth RNAi siRNA Negative Control Med GC Duplex (Cat. No. 12935300, 250 µL)
    - Stealth RNAi siRNA Negative Control Med GC Duplex #2 (Cat. No. 12935112, 250 µL)
    - Stealth RNAi siRNA Negative Control Hi GC Duplex #3 (Cat. No. 12935113, 250 µL)
    - Stealth RNAi siRNA Negative Control Low GC Duplex (Cat. No. 12935200, 250 µL)
    - Stealth RNAi siRNA Negative Control Low GC Duplex #2 (Cat. No. 12935110, 250 µL)
    - Stealth RNAi siRNA Negative Control Low GC Duplex #3 (Cat. No. 12935111, 250 µL)
    - Stealth RNAi siRNA Negative Control Kit (Cat. No. 12935100, contains one of each GC content level negative control, 250 µL each at 20 µM)
    - For a scrambled negative control, you could also scramble your designed siRNA or Stealth RNAi sequence using the BLOCK-iT RNAi Designer.

    You can also use our BLOCK-iT AlexaFluor Red Fluorescent Control (Cat. No. 14750100) or BLOCK-iT Fluorescent Oligo (Cat. No. 2013, 13750062) as a control for the transfection.

    How do I use Stealth Select? Should the Stealth Select 3 be transfected together as a pool or individually?

    Stealth Select sets are delivered as 3 separate vials containing each unique duplex. They are ready to transfect immediately once the lyophilized RNA is rehydrated at the desired concentration. We recommend that each sequence be transfected and tested individually, as pooling of multiple RNAi duplexes together increases the risk of negative off target effects in the cells, and further analysis is then needed to eliminate which sequences are contributing to the knockdown.

    How much Stealth siRNA do I need?

    The Stealth siRNA is delivered as 20 nmol of annealed duplex. It offers approximately 4000 transfections of 24-well size dishes (5 pmol/well). Invitrogen can synthesize selected sequences in larger scale upon request.

    How do I know the siRNA or Stealth RNAi will not target other genes?

    The RNAi Designer utilizes stringent design rules to select Stealth siRNA that are unique to the organism that you select. Stealth RNAi is most effective at minimizing this concern, as only the antisense strand is capable of eliciting the RNAi response.

    There are no pre-designed Stealth RNAi siRNA (Stealth Select) available for my gene. What can I do?

    If a pre-designed siRNA is unavailable, you can use our algorithm to design one using your target mRNA nucleotide sequence or accession number (species other than human, mouse, rate; 5' or 3' UTR). The RNAi Designer is a free online tool that is ideal for designing effective Stealth RNAi. The RNAi Designer uses a proprietary algorithm based on published design rules, sophisticated homology elimination, and mining of sequence information from Invitrogen's extensive wet lab data on successful RNAi. Using RNAi Designer, you'll be able to design a unique Stealth RNAi molecule to target your mRNA of interest for effective knockdown. If you have an existing siRNA sequence that works well for you, it is simple to change this to an effective Stealth duplex and get all the Stealth RNAi benefits.

    What is the difference between Stealth, Stealth Select, and Validated Stealth RNAi?

    Stealth RNAi is a 25-mer blunt dsRNA oligo that contains a chemically modified sense strand. While this modification is proprietary, it helps to:

    - Increase stability in the RISC complex
    - Increase stability in vitro
    - Decrease off-targeting effects of sense strand since it won't be used as the guiding strand
    Stealth RNAi can be ordered via web or email, and are available at 20 nmol, 80 nmol, 1 µmol, and 1.7 µmol. The RNAi is delivered lyophilized in annealing buffer, along with 1 mL DEPC water for dilution.

    Stealth Select RNAi are pre-designed Stealth RNAi molecules for mouse, rat or human. These sequences have been run through Smith-Waterman algorithm to further minimize off target effects. These sequences are mainly ORF targeted, but have not been bench-tested. This RNAi can be ordered online, and found on the RNAi Express Search engine. The top 3 non-overlapping targets per gene are provided, so that a set of three can be purchased. Sequences are provided on the CoA with delivered product. 20nmol of the annealed duplex will be delivered lyophilized, along with 1 mL DEPC water for dilution. These RNAi can also be ordered at 1 nmol through plate select. We guarantee 2 out of 3 Stealth Select RNAi siRNA targeted to the same gene will give you 70% or greater knockdown.

    Validated Stealth RNAi are predesigned and bench-tested Stealth RNAi. These RNAi are non-overlapping sequences targeting human genes. These RNAi are delivered at 20 nmol of each annealed duplex lyophilized, with 1 mL DEPC treated water. They can also be ordered at 1 nmol through plate select. These siRNA are guaranteed individually.

    What approach should be taken to measure target knockdown after siRNA transfection?

    Measure mRNA levels by real-time PCR. A typical time course analysis would measure at 24, 48, and 72 hours. Protein analysis may be checked at 48, 72, and 96 hours. This will vary depending on the gene being targeted.

    Ideally, how may siRNAs (against a particular gene) should be used while optimizing the experiment?

    We suggest using at least 2 siRNA targeting the same gene. This will give greater confidence in RNAi data.

    How can I know what sequence is responsible for an siRNA effect?

    In order to analyze the effects of a specific siRNA sequence on gene activity, the introduced siRNA sequence must be known. This requires the design, introduction, and measurement of gene blocking following the addition of synthetic siRNA oligonucleotides or of a short hairpin RNA (shRNA) sequence in a vector. When diced siRNAs (d-siRNA) are introduced into the cell, they are particularly effective at initiating an RNAi effect because generally there will be several effective siRNA sequences that are part of the pool. However, there currently there is no way to identify the specific sequence(s) in a pool that are responsible for the effect.

    How do I measure the effect of a siRNA?

    The most common way to measure gene specific knockdown is to perform real-time PCR. In some cases a reporter system that allows easy measurement of a reporter gene, such as beta-galactosidase, may be used. Western blot analysis to compare the level of protein expression before and after the introduction of siRNA may also be employed.

    What is the difference between siRNA and diced siRNA (d-siRNA)?

    The structure of the molecules is the same: Dicer specifically cleaves long dsRNA into the 21-23 nucleotide duplexes with a 2-nucleotide overhang that is the hallmark of siRNA. A key difference is that d-siRNA typically contains a pool of siRNA generated from the entire length of a long dsRNA target, whereas siRNA generally refers to a single sequence that is specific to a particular target region, and is often synthesized as a single oligo or a specified combination of several oligos.

    What are the ways that siRNA can be generated?

    The three most common methods of generating siRNA to introduce into mammalian cells are:

    - In vitro transcription and dicing
    - Synthetic siRNA
    - Vectors carrying an RNAi cassette

    What are the advantages of RNAi over other methods used for knocking down gene expression?

    RNAi is a cost-effective method for the rapid identification of gene function, and appears to work well for most genes tested to date. RNAi is rapidly becoming the preferred method for knocking out the expression of targeted genes. RNAi is useful for assigning gene function, signaling pathway analysis, RNAi mechanism studies, target validation, and shows tremendous potential for diagnostics and therapeutics.

    What is the difference between custom designed, custom synthesized, and custom modified siRNA?

    Custom Designed: If a pre-designed siRNA is unavailable, we will use our algorithm to custom design one for you using the target mRNA's nucleotide sequence or accession number (transcript variants, species other than human, mouse, rat). Custom designed siRNAs are not guaranteed.
    Custom Synthesized: The customer provides the siRNA sequence, and we synthesize it. Custom synthesized siRNAs are not guaranteed.
    Custom Modified: A quote can be requested for custom modifications such as fluorescent labels (FAM, Cy3), custom sizes ( greater than 50 nmol) and aliquotting. Custom modified siRNAs are not guaranteed.

    What does "off-targeting" mean?

    Off-targeting effects are when any gene-silencing effects are caused by siRNAs on nontarget mRNAs through the RNAi mechanism. They could come from the guide (antisense) or passenger (sense) strand of siRNA.

    Can the Stealth negative controls be used along with the Silencer Select experimental siRNAs and vice versa?

    Typically, we would recommend matching the experimental and control siRNAs by brand. There are differences in siRNA length and modification between Silencer, Stealth, and Silencer Select sequences. A good experimental design would minimize these variables between experimental and control siRNA. That said, in a bind, using a negative control with a different brand than the experimental siRNA would still help to control for off-target effects.

    Is there a way for me to screen sequences to determine if they work for RNAi knockdown?

    Yes, we offer a pSCREEN-iT/lacZ-DEST Gateway Vector Kit which can be used to assess the potency of any RNAi knockdown without knowing protein function, western blotting or qRT-PCR. The system uses a beta-galactosidase activity assay to measure the knockdown ability of an RNAi reagent (Stealth siRNA, Silencer siRNA, shRNA-containing plasmid, Dicer pools, etc). Clone your target gene into the vector provided, and co-transfect it along with the knockdown reagent being tested. Expressed genes will be fused to beta-galactosidase, enabling you to use beta-Gal levels to measure the amount of RNAi-induced degradation of the target gene.

    Why do I need a negative control for my siRNA experiment?

    A negative control is meant to reveal sequence-independent effects of siRNA on your cells. It should match the general chemistry of your positive molecule (length, modification) but be made up of a nontargeting sequence that will not target any specific gene.

    Do you offer negative and positive controls for my RNAi experiments?

    Yes, we offer several choices for negative and positive controls. Negative controls are typically universal nontargeting sequences, while positive controls are reporter genes that can be used as positive controls and/or protocol validation. Please select a control that has the same modification as your siRNA (i.e., Silencer Select Negative Control, Stealth Negative Control).

    What purification method should I choose when ordering siRNA?

    The purification method will depend on your experiment. Please see our general guidelines below:

    Standard purification:
    Desalted and analyzed by MALDI-TOF mass spectrometry
    Guaranteed to be at least 80% full-length product
    Recommended for adherent cell lines

    HPLC:
    HPLC purified and analyzed by MALDI-TOF and analytical HPLC
    Guaranteed to be at least 97% full-length product
    Recommended for electroporation of primary or suspension cell lines

    In vivo-ready:
    HPLC purified, analyzed by MALDI-TOF and analytical HPLC, dialyzed to remove salts, sterile filtered, and endotoxin tested
    Guaranteed to be at least 97% full-length product
    Recommended to researchers using siRNA in animals
    Please note that, in all cases, the efficiency of annealing is analyzed by PAGE.

    What is your siRNA guarantee?

    Please see the following:

    (a) Silencer Select siRNA: “Thermo Fisher Scientific guarantees that when you purchase two Silencer Select Pre-designed siRNA to the same target, then those two siRNAs will silence the target mRNA by 70% or more. To qualify for the guarantee, siRNAs must have been transfected at ?5 nM and mRNA levels detected 48 hours posttransfection. Real-time RT-PCR is recommended but not required for this application. Customers must also show sufficient knockdown with a positive control siRNA to demonstrate transfection efficiency. If the guaranteed level of knockdown is not observed and an appropriate positive control is successful, a new Silencer Select siRNA sequence will be synthesized free of charge. This guarantee does not extend to any replacement product.”
    (b) Stealth siRNA: “Thermo Fisher Scientific guarantees that when you purchase three Stealth Pre-designed siRNA to the same target, then at least two of those three independent, nonoverlapping siRNAs will silence the target mRNA by 70% or more. To qualify for the guarantee, siRNAs must have been transfected at ?20 nM and mRNA levels detected 48 hours posttransfection. Real-time RT-PCR is recommended but not required for this application. Customers must also show sufficient knockdown with a positive control siRNA to demonstrate transfection efficiency. If the guaranteed level of knockdown is not observed and an appropriate positive control is successful, a new Silencer siRNA sequence will be synthesized free of charge. This guarantee does not extend to any replacement product. We also recommend the use of an appropriate negative control, such as one of the three Stealth RNAi Negative Controls, to normalize message knockdown.”
    (c) Silencer siRNA: “Thermo Fisher Scientific guarantees that when you purchase three Silencer Pre-designed siRNAs to the same target, at least two of the siRNAs will reduce target mRNA levels in cultured cells by 70% or more when measured 48 hours after transfection at 100 nM or higher final siRNA concentration under the conditions described below. If at least two of the three siRNAs do not induce >70% target mRNA knockdown, Thermo Fisher Scientific will provide a one-time replacement of up to three Silencer Pre-designed siRNAs per target at no additional charge. Requests for replacement product must be made within one hundred and eighty (180) days from the date of delivery of the Silencer Pre-designed siRNAs. Optimum transfection efficiency must be confirmed using good laboratory practices and a proven-to-work siRNA to an endogenous message, such as Invitrogen Silencer GAPDH siRNA Control. To assess knockdown, target mRNA levels in treated samples must be compared to that of cells transfected with a nontargeting control siRNA, such as Silencer Negative Control #1. We recommend Applied Biosystems TaqMan Gene Expression Assays to quantify mRNA levels.”

    What types of controls are needed for a successful siRNA experiment?

    We recommend the following controls:

    - Positive control siRNA
    - Negative control siRNA
    - Cells-only control
    - Multiple siRNAs per target
    - Transfection reagent alone

    Can I transfect cells repeatedly to obtain sustained knockdown?

    Yes, if the cells are doing fine with the transfection protocol.

    How soon should I see knockdown after siRNA transfection?

    Depending on the gene you are working with, it can be measured at the mRNA level as soon as a few hours after transfection. We recommend assessing the mRNA knockdown 48 hours posttransfection. Factors affecting the timing include the transcription activity, the turnover rate for the messenger, and if there are alternative pathways. To determine the peak knockdown, it is best to perform a time course experiment.

    Which strand of the siRNA duplex is acting on the mRNA?

    The antisense (guide) strand will bind to the mRNA.

    What is siRNA?

    siRNA stands for “short interfering RNA”. It consists of two complementary RNA strands 19-21 nucleotides long, with TT or dTdT overhangs. Longer dsRNA will trigger increased immune responses and be degraded. The overhangs are thought to be added by dicer when the molecules are processed. Target cleavage is thought to be in the middle of anti-sense sequences, so the middle bases need to be conserved. The synthetics will be delivered into cells to initiate RNAi via electroporation or lipid delivery. Once delivered, the two strands will separate, releasing the antisense strand. With the aid of a protein, RISC, it binds to a complementary sense sequence on the molecule of mRNA. If the base-pairing is exact, the mRNA is destroyed.

    What is RNAi, and how does it work?

    RNAi stands for RNA interference. The molecules that mediate RNAi are short dsRNA oligonucleotides that are processed internally by an enzyme called Dicer. The Dicer cleavage products were first referred to as short interfering RNA, now known as siRNA. RNAi technology takes advantage of the cell's natural machinery to effectively knock down expression of a gene with transfected siRNA. There are several ways to induce RNAi: synthetic molecules, RNAi vectors, and in vitro dicing. In mammalian cells, short pieces of dsRNA - short interfering RNA- initiate the specific degradation of a targeted cellular mRNA. In this process, the antisense strand of siRNA becomes part of a multiprotein complex, or RNA-induced silencing complex (RISC), which then identifies the corresponding mRNA and cleaves it at a specific site. Next, this cleaved message is targeted for degradation, which ultimately results in the loss of protein expression.


    For Research Use Only. Not for use in diagnostic procedures.