Learn More
Myron L Company STD SOLUTION KCL70000 1 QUART
Catalog No. NC1602360
Encompass
KCl Standard Solution is used to calibrate conductivity instruments that read directly in microsiemens (micromhos) or millisiemens (1000 microsiemens). KCI (potassium chloride) is a very stable salt and is an international calibration standard for conductivity measurement.
Catalog No. NC1602360 Supplier Myron L Company Supplier No. KCL70000Q
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
KCl Standard Solution is used to calibrate conductivity instruments that read directly in microsiemens (micromhos) or millisiemens (1000 microsiemens). KCI (potassium chloride) is a very stable salt and is an international calibration standard for conductivity measurement.