missing translation for 'onlineSavingsMsg'
Learn More

LifeSign Status Mono Test Kit

Catalog No. 23264801
Encompass
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23264801 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23264801 Supplier LifeSign Supplier No. 68364

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Status Mono immunoassay for the qualitative detection of infectious mononucleosis heterophile antibodies in whole blood, serum or plasma

Features:

  • FDA cleared with no age restrictions
  • CLIA waived for whole blood
  • Single reagent procedure provides results in 3 to 8 minutes
  • Fingerstick whole blood convenience
  • 99.9% sensitivity for earlier IM detection
  • Available in multiple kit sizes; ideal for any practice size
  • New plastic sample transfer pipettes for increased safety

Specifications

Detectable Analytes Mononucleosis Antibodies
Format Kit
Sample Type Plasma, Serum, Whole Blood
Type Mononucleosis Immunoassay
Includes External QC negative and positive controls
Quantity 25 Tests
Specificity 0.988
Test Time 3 to 8 min.