Learn More
HemoCue America StatSpin™ Glass Microhematocrit Tubes
Catalog No. 22269245
Change view
Click to view available options
For Use With (Application):
Anti-coagulated samples
Heparinized for fingersticks
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
22269245 Heparinized for fingersticks
22269246 Anti-coagulated samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22269245 Supplier HemoCue America Supplier No. HT9H 10
Only null left
HemoCue America StatSpin Glass Microhematocrit Tubes, For Use With: Heparinized for fingersticks, RH12 HCT rotor, Microhematocrit Capillary Tube, Length: 1.57 in., 4cm, Recommended Applications: Healthcare, Shelf Life: 2 Year(s), Volume: 0.0003 oz., 9uL
Glass tubes are 40mm long.
Specifications
Product Type Microhematocrit Capillary Tube
Material Glass
Capacity (Metric) 9 μL
For Use With (Equipment) RH12 HCT Rotor
For Use With (Application) Heparinized for fingersticks
Length (Metric) 4 cm
Length (English) 1.57 in.