Learn More
StatLab StatPrep Preserve NonGyn
Non-gynecological cytology preservative.
Supplier: StatLab CY32NONGYN32
Alert:
Harmful if swallowed, in contact with skin, or if inhaled
Description
- 32 oz. bottle
- StatPrep™ preserve is used for the preservation and transport of cytology specimens intended for liquid-based cytology processing
Specifications
| Preservative | |
| Not Autoclavable | |
| Non-sterile | |
| No |