Learn More
StatLab StatPrep Cytology Fixative
Natural ethyl alcohol and polyethylene glycol formulation.
Supplier:  StatLab CYCFI4CS
Catalog No. STLCYCFI4CS
Only null left
Explore available promotions
Alert:
Harmful if swallowed, in contact with skin, or if inhaled
Description
Description
  • Economical, environmentally safe, and effective cell preservation fixative
  • 4 oz. spray bottle
  • 12 per case
Specifications
Specifications
All Tissue Processors
Fixative
Fixative
Liquid
Chemical Solution
4 oz.
Fixation
Solution
Product Suggestions
Product Suggestions