Learn More
StatLab™ Statmark™ Pen
Catalog No. STLSMPBK
Change view
Click to view available options
Color:
Black
Catalog No. Color
STLSMPBK Black
1 options
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STLSMPBK Supplier StatLab™ Supplier No. STLSMPBK
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Harmful if swallowed, if in contact with skin, or if inhaled.
Designed exclusively for lab techs, this original formulation does not dry out and withstands exposure to common processing chemicals. Black ink.
  • Great for both slides and embedding cassettes
Specifications
Tip Style Ultrafine
Product Type Permanent Lab Marker
Color Black
Autoclavable Not Autoclavable
Sterility Non-sterile
Disposable Single-use
Latex Free Yes
No. per Pack 12 Pk.
Packaging Type Box