missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Starter Sacrificial Inks for Matribot™ Bioprinter

Catalog No. 01257116
Change view
Click to view available options
Quantity:
10 Syringes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01257116 10 Syringes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257116 Supplier Corning™ Supplier No. 6159

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Starter Sacrificial Inks are sacrificial material and inks for educational purposes. Designed for Use with Matribot™ Bioprinter.

  • Water-soluble gel used as a sacrificial material and ink for educational purposes
  • Ideal for evaluating the printing of complex structures, prior to printing with human cells
  • Designed for use with Corning™ Matribot™ Bioprinter

Specifications

Color Transparent Blue
For Use With (Equipment) Corning Matribot Bioprinter
Product Type Sacrificial Ink
Capacity (Metric) 2.7 mL/Syringe
Description Sterile-A Quality
Model Starter
For Use With (Application) Evaluating the printing of complex structures, prior to printing with human cells
Quantity 10 Syringes
Storage Requirements 4°C to 8°C