missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Starter™ pH Meter ST2200
Description
- Equipped with a large 3.5 in. LCD display
- Features a 99 item memory for measurements and last calibration
- Features auto-buffer recognition and auto-endpoint modes that make measurement and calibration simple
- Choose between just the ST2200 meter with electrode holder and a full kit including ST2200 meter, electrode holder, ST320 electrode, 4 pH buffer mini kits (50 mL)
Specifications
Specifications
| Data Management Details | Datalog for Measurements and Last Calibration |
| Accuracy (pH) | ±0.01 |
| Accuracy (Temperature) | ±0.5°C |
| Accuracy (mV) | ±1.5 |
| Includes | ST2200 Meter, Electrode Holder |
| Measurement Modes | Auto-stop, Continuous |
| Calibration | Linear |
| Range (pH) | 0 to 14 |
| Resolution (pH) | 0.01 |
| Range (mV) | ±1999 |
| Show More |