Learn More
Starplex™ Scientific Specimen Containers: Cups
 View more versions of this product
Catalog No. 22146593
Only null left
Description
Description
  • Break- and leak-resistant for safety during harsh transport conditions or pneumatic tube systems
  • Clarified polypropylene cups provide easy visual analysis of sample
  • Full-circle thread design prevents cross-threading
  • Graduated markings allow quick measurement of sample
  • Labeling provides ample space for patient information
  • Ridges on cap provide a secure grip for safe handling
Specifications
Chemical Identifiers
Polypropylene
Specifications
Polypropylene
Non-sterile
90 mL
Wide mouth
Product Suggestions
Product Suggestions