Learn More
Starplex™ Scientific LeakBuster™ Specimen Containers: Sterile
Ideal for use in harsh transport conditions or in pneumatic tube systems
$444.61
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Color | Product Type | No. per Case | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Color | Product Type | No. per Case | Price | Quantity & Availability | ||
|
09735201
|
Starplex™ Scientific
NCS6021O |
60 mL | Orange | Specimen Container | 500/Cs. |
Case of 500 Each for $444.61
|
|
||
Description
Ideal for use in harsh transport conditions or in pneumatic tube systems
A premium line of containers designed for leakproof transportation.
- Streamlined straight-sided vials are stable, stackable, and save space in storage
- Caps are designed with a full-circle thread that prevents the cap from cross threading and leaking
- QuickTurn™ caps are provided on the wide-mouth containers; lids are secured with only a quarter of a turn
- Sterile containers have an orange cap and tamper tab patient identification label that meets current JCAHO standards
- LeakBuster containers have passed DOT/ICAO/IATA 95kPa Pressure Differential and Thermal Shock requirements
Order Info
Individual sterile containers have large identification labels and tamper-evident tabs.
Specifications
| Polypropylene | |
| Sterile | |
| Yes |