Learn More
Heidolph™ StarFish Workstation Component - PolyBlocks
Component parts used in the assembly of Heidolph Starfish self-designed, modular reaction stations
Supplier:  Heidolph™ 015890230
 View more versions of this product
Catalog No. 05404050
Only null left
Description
Description
  • Small, segment blocks (some with multiple wells) can be mixed and matched to accommodate various combinations of vessels
  • StarFish fits 5 PolyBlocks at once
  • Use with flasks, tubes, or vials
  • Anodized aluminum for strong chemical resistance and fast heat-up times up to 260 ºC
  • PolyBlocks are stackable to maximize lab space
  • PolyBlocks have a 3.5 mm hole for a temperature probe
  • PolyBlocks are individually labelled for easy identification
  • Optional reducing inserts for 250 mL block
Specifications
Chemical Identifiers
Polyblock
Specifications
9 x 15 mm Vial
Polyblock
Product Suggestions
Product Suggestions