Learn More
Heidolph™ StarFish Workstation Component - PolyBlocks
Component parts used in the assembly of Heidolph Starfish self-designed, modular reaction stations
Supplier: Heidolph™ 015890160
Description
- Small, segment blocks (some with multiple wells) can be mixed and matched to accommodate various combinations of vessels
- StarFish fits 5 PolyBlocks at once
- Use with flasks, tubes, or vials
- Anodized aluminum for strong chemical resistance and fast heat-up times up to 260 ºC
- PolyBlocks are stackable to maximize lab space
- PolyBlocks have a 3.5 mm hole for a temperature probe
- PolyBlocks are individually labelled for easy identification
- Optional reducing inserts for 250 mL block
Chemical Identifiers
| Polyblock |
Specifications
| 1 x 250 mL Flask | |
| Polyblock |