missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ StarFish™ Workstation Component: Base Plates

Catalog No. 13900176
Change view
Click to view available options
Diameter (Metric):
135 mm
145 mm
For Use With (Equipment):
Heidolph Hotplates
Radleys and IKA Hotplates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) For Use With (Equipment)
13900176 145 mm Heidolph Hotplates
05404028 135 mm Radleys and IKA Hotplates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13900176 Supplier Heidolph™ Supplier No. 015890020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Component part used in the assembly of Heidolph Radleys StarFish™ self-designed, modular reaction stations

  • Base plate fits securely onto the top plate maintaining good thermal contact with the heated surface
  • Handles available separately (Cat. No. 13-900-177)

Specifications

Diameter (English) 1.8 in.
Diameter (Metric) 145 mm
For Use With (Equipment) Heidolph Hotplates
For Use With (Application) To maintain good thermal contact with the heated surface
Type Base Plate