Learn More
METTLER TOLEDO™ STARe Software Silver Package
Description
- Five programs - one database - STARe software utilizes 5 main programs linked to one another via the database: Installation, Module Control, Method, Experiment, & Evaluation Window
- Unlimited evaluation possibilities - enormous flexibility
- Reliable automation - high sample throughput with result assessment dramatically improves efficiency
- Integrated database - guarantees the highest level of data security
- Modular concept - tailor-made solutions for current and future needs
- Easy and intuitive OneClick™ operation - saves time in training and in daily use
For use with 30421563, 30421564, 30319250, 30319252, 30421562, 30319254, 30293533, 30319177 & 30319178
Spécifications
Spécifications
| Description | STARe software |
| For Use With (Application) | METTLER TOLEDO DSC and/or TGA |
| For Use With (Equipment) | 30421563 |