missing translation for 'onlineSavingsMsg'
Learn More

Starch, pure, 1 wt.% aqueous solution; stabilized w. 100ppb HgI2

Catalog No. p-3213148
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC380641000 100 mL Plastic bottle
AC380645000 500 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC380641000 Supplier Thermo Scientific Chemicals Supplier No. 380641000
Only null left

CAS: 9005-84-9 | (C6H10O5)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Starch
Physical Form Solution
CAS 9005-84-9, 7732-18-5
Boiling Point 100.0°C
Molecular Formula (C6H10O5)n
MDL Number MFCD00082026
Density 1.0000g/mL
Quantity 100 mL
Grade Pure
Packaging Plastic bottle
Sensitivity Passes Test
Specific Gravity 1
Show More Show Less

RUO – Research Use Only