missing translation for 'onlineSavingsMsg'
Learn More

Starch Solution (Stabilized), Fisher Chemical™

Catalog No. SS4081
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SS4081 1 L Poly Bottle
SS4084 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SS4081 Supplier Thermo Fisher Scientific Supplier No. SS408 1
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For iodometry

Specifications

Color Colorless
Chemical Name or Material Starch Solution
Name Note Stabilized
Physical Form Liquid
CAS 9005-25-8, 64-19-7, 7732-18-5
Boiling Point 100°C
Melting Point 0°C
Molecular Formula (C6H10O5)n
MDL Number MFCD00082026
Density 1.0g/cm³
Quantity 1 L
Packaging Poly Bottle
Sensitivity Pass Test
Vapor Pressure 14mmHg
Show More Show Less