missing translation for 'onlineSavingsMsg'
Learn More

Starch Solution, Solstice

Catalog No. 6002398
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002398 100 mL
6002399 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002398 Supplier Solstice Supplier No. 319554100ML

Indicator, 1% in H2O

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications

Chemical Name or Material Starch Solution
Density 1.005g/mL at 25°C
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Quantity 100 mL