missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, ACS reagent, soluble
CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol
Supplier: Thermo Scientific Chemicals 424490020
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| White | |
| 5.0 to 7.0 (2% soln. at 25°C) | |
| Powder | |
| 256.0°C to 258.0°C | |
| MFCD00082026 | |
| Solubility in water: 50g/L (90°C). | |
| GUBGYTABKSRVRQ-ASMJPISFSA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol | |
| 439341 | |
| ACS Reagent | |
| Plastic bottle | |
| Passes Test |
| Starch | |
| Soluble | |
| 9005-84-9 | |
| C12H22O11 | |
| alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
| 2 kg | |
| C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O | |
| 342.297 | |
| CHEBI:18167 | |
| Authentic | |
| 0.4% max. |
Chemical Identifiers
| 9005-84-9 | |
| 342.297 | |
| GUBGYTABKSRVRQ-ASMJPISFSA-N | |
| 439341 | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol |
| C12H22O11 | |
| MFCD00082026 | |
| alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose | |
| CHEBI:18167 | |
| C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O |
Safety and Handling
EINECSNumber : 232-686-4
TSCA : TSCA
RUO – Research Use Only