missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, Soluble, ACS Reagent, Reagents
Specifications
Specifications
| Boiling Range | 668°C |
| Color | White |
| Boiling Point | 668°C |
| Packaging | Glass Bottle |
| Quantity | 125 g |
| Specific Gravity | 1.5 |
| Grade | ACS |
| Physical Form | Solid |
| Chemical Name or Material | Starch, Soluble, ACS Reagent |